missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Texwipe™ Revolve™ Sealed Edge Pre-Wetted Wipers
Made from 100% upcycled polyester material and processed on Texwipe’s fully automated manufacturing system. Revolve™ Wipers contribute to sustainability goals by reducing the amount of post-consumer water bottles going into landfills and reducing carbon emission levels.
Brand: Texwipe™ STX1704P
This item is not returnable.
View return policy
Description
- By using Revolve™ products, companies participate in the reuse of polyester polymers, saving the number of bottles indicated on each product bag from going into landfill
- Revolve™ processing provides low levels of ions, NVRs (non-volatile residues), particles and fibers for use in critical cleaning applications and environments
- Designed for high sorption capacity which is ideal for spill control, cleaning, and solution applications
- Meets USP <797> and USP <800> wiper requirements
- Pre-wet with 0.2 μm filtered USP-grade 70% IPA / 30% DI Water
- Wipers provide consistent, optimized cleaning efficiency with repeatable wetness and VOC levels
- Packaged in easy-to-use, recloseable slider bags that reduce solution evaporation to preserve a consistent wiper wetness level
- Sterile wipers are triple-bagged with a case liner forming a forth layer of protection
- Gamma irradiated to a Sterility Assurance Level of 10-6 according to AAMI Guidelines
- Individually lot coded for traceability and quality control
- Sterile Validation Documentation available
Specifications
Cleanroom Wiper | |
10 cm | |
10 cm | |
Sealed | |
Cleaning, Applying and Removing Disinfectants | |
Sterile |
4 in. | |
4 in. | |
Polyester | |
4 x 4 in. | |
70% IPA/ 30% DI Water | |
Made From 100% Upcycled Polyester Material |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- X200 Revolve 10X10CM Sterile Wipers Pre-Wetted Wit h 70% IPA,50 wipers/bag
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : isopropyl alcohol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction