missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REXO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15598-1-AP-150UL
This item is not returnable.
View return policy
Description
Rex2 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.Specifications
REXO2 | |
Polyclonal | |
Unconjugated | |
REXO2 | |
CGI 114, REX2, REXO2, RFN, RNA exonuclease 2 homolog, SFN, Small fragment nuclease, SMFN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100043034, 100362123, 25996 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y3B8 | |
Rex2, REXO2 | |
REXO2 Fusion Protein Ag7976 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |