Learn More
Tocris Bioscience™ Rf470DL
Blue rotor-fluorogenic fluorescent D-amino acid for labeling peptidoglycans in live bacteria
Brand:  Tocris Bioscience™ 7406/2
Additional Details : Weight : 0.00970kg
Product Code. 18759796
  • £323.83 / 2mg
Estimated Shipment: 29-07-2024
to see stock.
Description
Description
Rf470DL is a blue rotor-fluorogenic fluorescent D-amino acid for labeling peptidoglycans in live bacteria. Rf470DL fluoresces only when it becomes incorporated into the peptidoglycan, enabling no wash experiments. Excitation/emission λ ∽470/620 nm; quantum yield = 0.042; extinction coefficient 33,106 M-1 cm-1 (measured in PBS containing 50% glycerol).
Specifications
Specifications
N6-[3-[6-[5-[2-Carboxy-2-cyanoethenyl]-2-thienyl]-3,4-dihydro-1(2 H)-quinolinyl]-1-oxopropyl]-D-lysine hydrochloride
Fluorescent Probes for Imaging Bacteria
0.95
2 mg
C26H30N4O5S.HCl