Learn More
RGX-104, MedChemExpress
MedChemExpress RGX-104 is an orally bioavailable and potent liver-X nuclear hormone receptor (LXR) agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
£125.84 - £1430.22
Specifications
Chemical Name or Material | RGX-104 |
---|---|
CAS | 610318-54-2 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C34H33ClF3NO3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16495346
|
Medchem Express
HY-111498A |
100 mg |
£1430.22
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16487197
|
Medchem Express
HY-111498A |
5 mg |
£194.81
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16470787
|
Medchem Express
HY-111498A |
50 mg |
£960.74
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16460136
|
Medchem Express
HY-111498A |
2 mg |
£125.84
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16444478
|
Medchem Express
HY-111498A |
10 mg |
£308.55
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16443106
|
Medchem Express
HY-111498A |
25 mg |
£618.31
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RGX-104 is an orally bioavailable and potent liver-X nuclear hormone receptor (LXR) agonist that modulates innate immunity via transcriptional activation of the ApoE gene.Specifications
RGX-104 | |
White | |
C34H33ClF3NO3 | |
O=C(O)CC1=CC=CC(OCC[C@H](N(CC2=CC=CC(C(F)(F)F)=C2Cl)CC(C3=CC=CC=C3)C4=CC=CC=C4)C)=C1 | |
596.08 | |
Research | |
Cancer-programmed cell death |
610318-54-2 | |
Research | |
DMSO : 100 mg/mL (167.76 mM; Need ultrasonic) | |
596.08 | |
99.35% | |
Solid |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Recommended Storage : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.