Learn More
Thermo Scientific™ RheBuild™ Kits
Everything you need to maintain your Thermo Scientific™ Rheodyne™ valves and injectors in your HPLC system.
Brand: Thermo Scientific™ 7725-999
Code : A0
Additional Details : Weight : 0.20000kg
Description
Use Thermo Scientific™ RheBuild Kits to maintain your Thermo Scientific™ Rheodyne™ valves and injectors for high-performance liquid chromatography (HPLC). A single kit includes everything you need.
Spécification
Maintenance Kit | |
For Rheodyne models 7725/7725 i/7726 | |
Rheodyne Injector Models 7725/7725i/7726 |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.