missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhenium foil, 1.0mm (0.04in) thick, 99.97% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 x 25 mm
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 7440-15-5 |
Molecular Formula | Re |
MDL Number | MFCD00011195 |
Quantity | 25 x 25 mm |
Synonym | rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets |
InChI Key | WUAPFZMCVAUBPE-UHFFFAOYSA-N |
SMILES | [Re] |
IUPAC Name | rhenium |
Molecular Weight (g/mol) | 186.21 |
PubChem CID | 23947 |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Rhenium foil, 1.0mm (0.04in) thick, 99.97% (metals basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction