missing translation for 'onlineSavingsMsg'
Learn More
Rhenium(IV) sulfide, 99%, Thermo Scientific Chemicals Product Code.: 11423790

Rhenium(IV) sulfide, 99%, Thermo Scientific Chemicals

Product Code. 11423790
10 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 11423790

Brand: Thermo Scientific Alfa Aesar 089482.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rhenium(IV) sulfide is used as a diamagnetic semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12038-63-0
Molecular Formula ReS2
Molecular Weight (g/mol) 250.33
MDL Number MFCD00016307
InChI Key USWJSZNKYVUTIE-UHFFFAOYSA-N
Synonym rhenium sulfide, rhenium disulphide, disulfanylidenerhenium, dithioxorhenium, rhenium iv sulfide, bis sulfanylidene rhenium
PubChem CID 82864
IUPAC Name bis(sulfanylidene)rhenium
SMILES S=[Re]=S

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water. Soluble in 1N sodium hydroxide solution.
Formula Weight 250.33
Percent Purity 99%
Odor Odorless
Density 7.506 g/mL
Chemical Name or Material Rhenium (IV) sulfide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR10H032 Certificate TypeCertificate of Analysis Date30/07/2024 Catalog Number089482.04,089482.09
Lot NumberZ12E045 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number089482.04,089482.09
Lot NumberP05E010 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number089482.04,089482.09
Lot NumberL23N31 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number089482.04,089482.09
Lot NumberH14R026 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number089482.04,089482.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhenium(IV) sulfide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.