Learn More
Rhenium standard solution, for AAS, 1mg/mL Re in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 196221000
Code : 29
Detalles adicionales : CAS Number : 7440-15-5 Weight : 0.09960kg
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopyEspecificaciones
Rhenium standard solution | |
13598-65-7,7732-18-5 | |
100 mL | |
H4NO4Re | |
XPSXHXWHJAIQJR-UHFFFAOYSA-M | |
rheniumoylol amine | |
3084163 | |
Miscible with water |
997 to 1003 μg/mL of Re (4+) in water (20°C) | |
Plastic bottle | |
Colorless | |
MFCD00012377 | |
N.O[Re](=O)(=O)=O | |
268.24 | |
AAS | |
Liquid |
RUO – Research Use Only