missing translation for 'onlineSavingsMsg'
Learn More
Rhenium(VII) oxide, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11388428

Rhenium(VII) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11388428
50 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11388428

Brand: Thermo Scientific Alfa Aesar 040468.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

As a catalyst for oxidation/reduction reactions

As a catalyst for oxidation/reduction reactionsRhenium(VII) oxide acts as a precursor to methylrhenium trioxide, organorhenium oxides which catalyze olefin oxidation, metathesis and other transformations. It is used as a reagent for dehydrations and rearrangements of allylic alcohols, oximes and amides, oxidative cyclizations, alkene metathesis and formation of bisperoxyacetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-68-7
Molecular Formula O7Re2
Molecular Weight (g/mol) 484.407
MDL Number MFCD00011199
InChI Key NBGOSNNAAHDRLK-UHFFFAOYSA-N
Synonym rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basisShow MoreShow Less
PubChem CID 123106
IUPAC Name trioxo(trioxorheniooxy)rhenium
SMILES O=[Re](=O)(=O)O[Re](=O)(=O)=O

Specifications

Boiling Point 360°C
Melting Point 297°C
Color Green to Yellow
Physical Form Crystalline
Quantity 50 g
Linear Formula Re2O7
UN Number UN3085
Merck Index 14,8178
Solubility Information Dissolves in water to form HReO4; Soluble in alkalis and acids; Soluble in water and alcohol.
Formula Weight 484.4
Percent Purity 99.9%
Odor Odorless
Packaging Ampule
Sensitivity Moisture sensitive
Density 6.103 g/mL
Assay (metals basis)
Chemical Name or Material Rhenium(VII) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT03D026 Certificate TypeCertificate of Analysis Date27/12/2023 Catalog Number040468.09,040468.04,040468.18
Lot Number10248939 Certificate TypeCertificate of Analysis Date08/12/2023 Catalog Number040468.09,040468.04,040468.18
Lot NumberG20S044 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040468.09,040468.04,040468.18
Lot NumberX16B003 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040468.09,040468.04,040468.18
Lot NumberM19H005 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040468.09,040468.04,040468.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhenium(VII) oxide, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.