Learn More
Rhenium(VII) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Description
As a catalyst for oxidation/reduction reactionsRhenium(VII) oxide acts as a precursor to methylrhenium trioxide, organorhenium oxides which catalyze olefin oxidation, metathesis and other transformations. It is used as a reagent for dehydrations and rearrangements of allylic alcohols, oximes and amides, oxidative cyclizations, alkene metathesis and formation of bisperoxyacetals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon

Specifications
Specifications
Boiling Point | 360°C |
Melting Point | 297°C |
Color | Green to Yellow |
Physical Form | Crystalline |
Quantity | 10 g |
Linear Formula | Re2O7 |
UN Number | UN3085 |
Merck Index | 14,8178 |
Solubility Information | Dissolves in water to form HReO4; Soluble in alkalis and acids; Soluble in water and alcohol. |
Formula Weight | 484.4 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T03D026 | Certificate of Analysis | 27/12/2023 | 040468.09,040468.04,040468.18 |
10248939 | Certificate of Analysis | 08/12/2023 | 040468.09,040468.04,040468.18 |
G20S044 | Certificate of Analysis | 08/12/2021 | 040468.09,040468.04,040468.18 |
X16B003 | Certificate of Analysis | 08/12/2021 | 040468.09,040468.04,040468.18 |
M19H005 | Certificate of Analysis | 08/12/2021 | 040468.09,040468.04,040468.18 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.