missing translation for 'onlineSavingsMsg'
Learn More
Rhenium(VII) oxide, 99.995% (metals basis), Thermo Scientific Chemicals Product Code.: 15412347

Rhenium(VII) oxide, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 15412347
2 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 15412347

Brand: Thermo Scientific Alfa Aesar 045538.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rhenium(VII) oxide is a precursor to organorhenium oxides which catalyze olefin oxidation, metathesis and other transformations. It is also used as a catalyst for hemiacetal substitution and as an oxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-68-7
Molecular Formula O7Re2
Molecular Weight (g/mol) 484.407
MDL Number MFCD00011199
InChI Key NBGOSNNAAHDRLK-UHFFFAOYSA-N
Synonym rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basis
PubChem CID 123106
IUPAC Name trioxo(trioxorheniooxy)rhenium
SMILES O=[Re](=O)(=O)O[Re](=O)(=O)=O

Specifications

Boiling Point 360°C
Melting Point 297°C
Color Yellow
Physical Form Crystalline
Quantity 2 g
Solubility Information Soluble in water and alcohol.
Formula Weight 484.4
Odor Odorless
Sensitivity Moisture sensitive
Density 6.103
Chemical Name or Material Rhenium(VII) oxide
Show More Show Less
compliance-icons
Product Identifier
  • Rhenium(VII) oxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Rhenium oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhenium(VII) oxide, 99.995% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.