missing translation for 'onlineSavingsMsg'
Learn More

Rhenium wire, 0.25mm (0.01in) dia, 99.97% (metals basis), Thermo Scientific Chemicals

£44.72 - £620.09

Chemical Identifiers

CAS 7440-15-5
Molecular Formula Re
Molecular Weight (g/mol) 186.21
MDL Number MFCD00011195
InChI Key WUAPFZMCVAUBPE-UHFFFAOYSA-N
Synonym rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
PubChem CID 23947
ChEBI CHEBI:49882
IUPAC Name rhenium
SMILES [Re]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11341408
View Documents
Thermo Scientific Alfa Aesar
010311.BS
10 cm
£44.72
10cm
Please to purchase this item. Need a web account? Register with us today!
11361408
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
010311.BW
50 cm
£160.80
50cm
Please to purchase this item. Need a web account? Register with us today!
11351408
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
010311.CB
250 cm
£620.09
250cm
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation

Chemische Identifikatoren

7440-15-5
186.21
WUAPFZMCVAUBPE-UHFFFAOYSA-N
23947
rhenium
Re
MFCD00011195
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
CHEBI:49882
[Re]

Spezifikation

7440-15-5
MFCD00011195
WUAPFZMCVAUBPE-UHFFFAOYSA-N
rhenium
23947
99.97%
≈1.03g/m
(metals basis)
Re
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
[Re]
186.21
CHEBI:49882
Odorless
Wire
Rhenium wire, 0.25mm (0.01 in.) dia.
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

missing translation for 'einecsNumber' : 231-124-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate

RUO – Research Use Only