Learn More
Rhenium wire, 0.5mm (0.02in) dia, 99.97% (metals basis), Thermo Scientific Chemicals
Form: Approx.4.13g/m
£48.24 - £523.61
Chemical Identifiers
CAS | 7440-15-5 |
---|---|
Molecular Formula | Re |
Molecular Weight (g/mol) | 186.21 |
MDL Number | MFCD00011195 |
InChI Key | WUAPFZMCVAUBPE-UHFFFAOYSA-N |
Synonym | rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets |
PubChem CID | 23947 |
ChEBI | CHEBI:49882 |
IUPAC Name | rhenium |
SMILES | [Re] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11371408
|
Thermo Scientific Alfa Aesar
010312.BQ |
5 cm |
£48.24
5cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391408
|
Thermo Scientific Alfa Aesar
010312.BU |
25 cm |
£174.87
25cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381408
|
Thermo Scientific Alfa Aesar
010312.BY |
100 cm |
£523.61
100cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-15-5 | |
186.21 | |
WUAPFZMCVAUBPE-UHFFFAOYSA-N | |
23947 | |
rhenium |
Re | |
MFCD00011195 | |
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pelletsShow More | |
CHEBI:49882 | |
[Re] |
Specifications
Silver to Gray | |
Silver gray wire | |
Re | |
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets | |
[Re] | |
186.21 | |
CHEBI:49882 | |
Wire |
7440-15-5 | |
99.97% (Metals basis) | |
MFCD00011195 | |
WUAPFZMCVAUBPE-UHFFFAOYSA-N | |
rhenium | |
23947 | |
Odorless | |
Rhenium |
Safety and Handling
missing translation for 'einecsNumber' : 231-124-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.