Learn More
Thermo Scientific™ Rheodyne™ Suction Needle Adapter
Shop All Thermo Scientific ProductsDescription
Use the Thermo Scientific™ Rheodyne™ Suction Needle Adapter for suction loading of samples into the Rheodyne 9725 and 9725i Sample Injectors for high-performance liquid chromatography (HPLC),
Specifications
Specifications
| Type | Suction Needle Adapter |
| For Use With (Application) | Liquid Chromatography |
| Description | Suction Needle Adapter |
| For Use With (Equipment) | Rheodyne Injector Models 9725 and 9725i |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.