missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Rhodamine 6G, 99%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 290.0°C |
Assay Percent Range | 99% |
Beilstein | 18, V,12, 283 |
Solubility Information | Solubility in water: soluble. |
Formula Weight | 479.01 |
Percent Purity | 99% |
Grade | Pure |
Packaging | Glass bottle |
Quantity | 1 g |
Chemical Name or Material | Rhodamine 6G |
Safety and Handling
Product Identifier
- Rhodamine 6G
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'