Learn More
Thermo Scientific Chemicals Rhodamine 6G 99%
£21.00 - £62.90
Chemical Identifiers
CAS | 989-38-8 |
---|---|
Molecular Formula | C28H31ClN2O3 |
Molecular Weight (g/mol) | 479.017 |
MDL Number | MFCD00012665 |
InChI Key | VYXSBFYARXAAKO-UHFFFAOYSA-N |
Synonym | Basic Red 1, C.I. 45160 |
PubChem CID | 51358423 |
IUPAC Name | ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride |
SMILES | [H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10147990
|
Thermo Scientific Chemicals
419010050 |
5 g | Glass bottle |
£21.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10045820
|
Thermo Scientific Chemicals
419010250 |
25 g | Glass bottle |
£29.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10587362
|
Thermo Scientific Chemicals
419011000 |
100 g | Glass bottle |
£62.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
989-38-8 | |
479.017 | |
VYXSBFYARXAAKO-UHFFFAOYSA-N | |
51358423 | |
[H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-] |
C28H31ClN2O3 | |
MFCD00012665 | |
Basic Red 1, C.I. 45160 | |
ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride |
Specifications
290.0°C | |
99% | |
MFCD00012665 | |
Basic Red 1, C.I. 45160 | |
VYXSBFYARXAAKO-UHFFFAOYSA-N | |
[H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-] | |
479.017 | |
479.01 | |
Pure | |
Brown to Brown/Red | |
Rhodamine 6G, 99% |
989-38-8 | |
C28H31ClN2O3 | |
18, V,12, 283 | |
Solubility in water: soluble. | |
Authentic | |
ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride | |
51358423 | |
99% | |
98.5% min. | |
Fine Crystalline Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 213-584-9
missing translation for 'rtecsNumber' : DH0175000
missing translation for 'tsca' : TSCA
- Rhodamine 6G
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only