Learn More
Thermo Scientific Chemicals Rhodamine B, 98+%
£21.70 - £251.00
Chemical Identifiers
CAS | 81-88-9 |
---|---|
Molecular Formula | C28H31ClN2O3 |
Molecular Weight (g/mol) | 479.02 |
MDL Number | MFCD00011931 |
InChI Key | PYWVYCXTNDRMGF-UHFFFAOYSA-N |
Synonym | Basic Violet 10, C.I. 45170 |
PubChem CID | 6694 |
ChEBI | CHEBI:52334 |
IUPAC Name | [9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride |
SMILES | [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10440341
|
Thermo Scientific Chemicals
296570100 |
10 g | Glass bottle |
£21.70
10g |
In Stock
Log in to see stock. |
||||
10093370
|
Thermo Scientific Chemicals
296570250 |
25 g | Glass bottle |
£23.50
25g |
In Stock
Log in to see stock. |
||||
10035463
|
Thermo Scientific Chemicals
296571000 |
100 g | Glass bottle |
£37.90
100g |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
13428498
|
Thermo Scientific Chemicals
296570010 |
1 kg | Plastic Drum |
£251.00
1kg |
Estimated Shipment: 17-10-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
81-88-9 | |
479.02 | |
PYWVYCXTNDRMGF-UHFFFAOYSA-N | |
6694 | |
[9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride |
C28H31ClN2O3 | |
MFCD00011931 | |
Basic Violet 10, C.I. 45170 | |
CHEBI:52334 | |
[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC |
Specifications
210.0°C to 211.0°C | |
≥98% | |
MFCD00011931 | |
15, 8307 | |
Solubility in water: soluble. Other solubilities: slightly soluble in hcl and naoh, soluble in alcohol | |
Authentic | |
[9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride | |
6694 | |
479.01 | |
Pure | |
>255°C | |
Powder |
81-88-9 | |
C28H31ClN2O3 | |
19, V,8, 669 | |
Basic Violet 10, C.I. 45170 | |
PYWVYCXTNDRMGF-UHFFFAOYSA-N | |
[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC | |
479.02 | |
CHEBI:52334 | |
≥98% | |
Vapor Pressure: Negligible | |
Green | |
Rhodamine B, Pure |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Suspected of causing genetic defects.
Harmful if swallowed.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Rhodamine B
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only