missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Rhodamine Red™ C2 Maleimide
Rhodamine Red™ C2 maleimide
Brand: Molecular Probes™ R6029
253.84 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
The thiol-reactive Rhodamine Red™ C2 maleimide can be used to create red-fluorescent bioconjugates with excitation/emission maxima ∼560/580 nm.Specifications
5 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Rhodamine Red™, Rhodamine Red™ C2 maleimide | |
Rhodamine Red™ | |
560 |
Maleimide | |
Thiol | |
Maleimide | |
Classic Dyes | |
580 |
For Research Use Only. Not for use in diagnostic procedures.