missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Rhodamine WT, 20% solution in water
£44.40 - £251.00
Especificaciones
Chemical Name or Material | Rhodamine WT |
---|---|
CAS | 37299-86-8, 7732-18-5 |
Molecular Formula | C29H29ClN2Na2O5 |
Solubility Information | Solubility in water: miscible. |
InChI Key | NRZDMKVYRRMFRR-UHFFFAOYSA-L |
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Rhodamine WT | |
C29H29ClN2Na2O5 | |
NRZDMKVYRRMFRR-UHFFFAOYSA-L | |
disodium;4-[3-(diethylamino)-6-diethylazaniumylidenexanthen-9-yl]benzene-1,3-dicarboxylate;chloride | |
37718 | |
Glass bottle | |
1.1600g/mL |
37299-86-8, 7732-18-5 | |
Solubility in water: miscible. | |
CCN(CC)C1=CC2=C(C=C1)C(=C3C=CC(=[N+](CC)CC)C=C3O2)C4=C(C=C(C=C4)C(=O)[O-])C(=O)[O-].[Na+].[Na+].[Cl-] | |
566.99 | |
566.99 | |
1.16 |
Sicurezza e movimentazione
Identificativo prodotto
- Rhodamine WT, 20% solution in water
RUO – Research Use Only