Learn More
Rhodium, 5% on alumina powder, C301099-5, Thermo Scientific Chemicals
Description
Rhodium is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Rhodium is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with organic materials and flammable substances.

Specifications
Specifications
Color | Gray |
Physical Form | Powder |
Quantity | 25 g |
Solubility Information | Insoluble in water. |
Odor | Odorless |
Chemical Name or Material | Rhodium, 5% on alumina powder, C301099-5 |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P31I027 | Certificate of Analysis | 12/04/2022 | H36201.14,H36201.06,H36201.22 |
10196276 | Certificate of Analysis | 07/12/2021 | H36201.14,H36201.06,H36201.22 |
Z21G040 | Certificate of Analysis | 07/12/2021 | H36201.14,H36201.06,H36201.22 |
W26F008 | Certificate of Analysis | 07/12/2021 | H36201.14,H36201.06,H36201.22 |
41177002 | Certificate of Analysis | 07/12/2021 | H36201.14,H36201.06,H36201.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.