missing translation for 'onlineSavingsMsg'
Learn More

Rhodium, 5% on alumina powder, reduced, Thermo Scientific™

POA

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11311838
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
011770.04
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11391828
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
011770.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7440-16-6
102.91
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD00011201 MFCD03458392
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
CHEBI:33359
[Rh]

Specifications

Black
Black powder
MFCD00011201 MFCD03458392
Insoluble in water.
[Rh]
102.91
CHEBI:33359
Rhodium
7440-16-6
Rh
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
23948
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-125-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.