missing translation for 'onlineSavingsMsg'
Learn More

Rhodium on activated carbon, 5% Rh, unreduced, dry, Thermo Scientific Chemicals

£121.61 - £2619.03

Chemical Identifiers

CAS 7440-16-6, 7440-44-0
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10754733
View Documents
Thermo Scientific Acros
195440010
1 g Glass bottle
£121.61
1g
Please to purchase this item. Need a web account? Register with us today!
10378140
View Documents Promotion Details
Thermo Scientific Acros
195440050
5 g Glass bottle
£605.01
5g
Please to purchase this item. Need a web account? Register with us today!
10511261
View Documents
Thermo Scientific Acros
195440250
25 g Glass bottle
£2619.03
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-16-6, 7440-44-0
102.91
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD00011201 MFCD03458392
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
CHEBI:33359
[Rh]

Specifications

Black to Black-Gray
Rh
01,982; 03,326; 04,418; 06,503; 08,433
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
[Rh]
102.91
CHEBI:33359
5% (Rh)
Powder
7440-16-6, 7440-44-0
MFCD00011201 MFCD03458392
15, 8310
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
23948
102.91
Typical particle size distribution: D50: 20 Micron, <8 Micron: 17%, <16 Micron: 39%,< 32 Micron: 67%, <64 Micron: 90%, <128 Micron: 99%
Rhodium on activated carbon, 5% Rh
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only