missing translation for 'onlineSavingsMsg'
Learn More
Rhodium black, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 15411017

Rhodium black, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 15411017
2 g
Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy

Product Code. 15411017

Brand: Thermo Scientific Alfa Aesar 012353.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]

Specifications

Color Gray
Appearance gray powder
Assay Percent Range 99.9% (Metals basis)
Quantity 2 g
Odor Odorless
Form Powder
Chemical Name or Material Rhodium black

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX20I022 Certificate TypeCertificate of Analysis Date26/10/2022 Catalog Number012353.04,012353.02
Lot NumberS24I055 Certificate TypeCertificate of Analysis Date25/07/2022 Catalog Number012353.04,012353.02
Lot NumberS21C028 Certificate TypeCertificate of Analysis Date18/04/2022 Catalog Number012353.04,012353.02
Lot NumberL11N23 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012353.04,012353.02
Lot NumberF07K23 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012353.04,012353.02

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhodium black, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.