missing translation for 'onlineSavingsMsg'
Learn More
Rhodium(II) acetate dimer, anhydrous, ca 46% Rh, Thermo Scientific Chemicals Product Code.: 10083250

Rhodium(II) acetate dimer, anhydrous, ca 46% Rh, Thermo Scientific Chemicals

Product Code. 10083250
1 g, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10083250

Brand: Thermo Scientific Acros 268630010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15956-28-2
Molecular Formula C8H12O8Rh2
Molecular Weight (g/mol) 441.99
MDL Number MFCD00003538
InChI Key VOXKIADTKKPBBP-UHFFFAOYSA-N
Synonym rhodium ii acetate, rhodium diacetate, acetic acid, rhodium 2+ salt, rhodium ii acetate dimer dihydrate, dirhodium tetraacetate, rhodium ii acetate dimer, rhodium acetate, brown, water soluble, diacetoxyrhodiumShow MoreShow Less
PubChem CID 122130469
SMILES C[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4

Specifications

Melting Point 205.0°C
Color Green
Physical Form Micro-crystalline Powder
Quantity 1 g
Merck Index 15,8311
Solubility Information Solubility in water: insoluble. Other solubilities: moderately soluble in ethanol, acetone, acetonitri, le, dmso, thf
IUPAC Name rhodium(2+);tetraacetate
Formula Weight 441.98
Percent Purity ∼46% (Rh)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Rhodium(II) acetate dimer
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0433806 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog Number268632500,268630010,268630050
Lot NumberA0448636 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number268632500,268630010,268630050
Lot NumberA0401536 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number268632500,268630010,268630050
Lot NumberA0397015 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number268632500,268630010,268630050
Lot NumberA0409522 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number268632500,268630010,268630050
compliance-icons
Product Identifier
  • Rhodium(II) acetate dimer, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhodium(II) acetate dimer, anhydrous, ca 46% Rh, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.