missing translation for 'onlineSavingsMsg'
Learn More
Rhodium(II) acetate, dimer, Premion™, 99.99% (metals basis), Rh 46.2% min, Thermo Scientific Chemicals Product Code.: 11370709

Rhodium(II) acetate, dimer, Premion™, 99.99% (metals basis), Rh 46.2% min, Thermo Scientific Chemicals

Product Code. 11370709
1 g
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy

Product Code. 11370709

Brand: Thermo Scientific Alfa Aesar 010560.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use in Cyclopropanation of Alkenes

Catalyst for cyclopropanation of alkenes, oxidation of alcohols, cyclization reactions involving -diazo carbonyl groups, insertion into C-H and X-H bonds (X-H is NH or SH or OH) and for ylide formation. More reactive and useful in differentiating ribonucleosides and deoxynucleosides. It is also used in functionalizing fullerenes into polymers. Efficient catalyst for hydrogen transfer from 2-propanol to cyclohexanone and other unsaturated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15956-28-2
Molecular Formula C8H12O8Rh2
Molecular Weight (g/mol) 441.99
MDL Number MFCD00003538
InChI Key VOXKIADTKKPBBP-UHFFFAOYSA-N
Synonym rhodium ii acetate, rhodium diacetate, acetic acid, rhodium 2+ salt, rhodium ii acetate dimer dihydrate, dirhodium tetraacetate, rhodium ii acetate dimer, rhodium acetate, brown, water soluble, diacetoxyrhodiumShow MoreShow Less
PubChem CID 122130469
SMILES C[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4

Specifications

Melting Point 205°C
Color Green
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in aqueous solution like water. Slightly soluble in ethanol
IUPAC Name tetramethyl-2,4,6,8,9,11,12,14-octaoxa-1,5-dirhodatetracyclo[3.3.3.3¹,⁵.0¹,⁵]tetradecane-1,1,5,5-tetrakis(ylium)-3,7,10,13-tetraide
Formula Weight 441.99
Odor Odorless
Appearance Green powder
Chemical Name or Material Rhodium(II) acetate, dimer, Rh 46.2% min.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW22J035 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog Number010560.01,010560.03,010560.06
Lot NumberW08I004 Certificate TypeCertificate of Analysis Date14/09/2022 Catalog Number010561.02,010560.01,010560.03,010560.06
Lot NumberJ31N28 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010561.02,010560.01,010560.03,010560.06
Lot NumberQ09A043 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010561.02,010560.01,010560.03,010560.06
Lot NumberL07Q023 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010561.02,010560.01,010560.03,010560.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhodium(II) acetate, dimer, Premion™, 99.99% (metals basis), Rh 46.2% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.