missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(II) trifluoroacetate dimer

Product Code. p-3586491 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10568833

Brand: Thermo Scientific Acros 389702500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31126-95-1 | C8F12O8Rh2 | 657.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31126-95-1
Molecular Formula C8F12O8Rh2
Molecular Weight (g/mol) 657.88
MDL Number MFCD00209611
InChI Key SZQVMUPTZFMHQT-UHFFFAOYSA-N
Synonym rhodium ii trifluoroacetate dimer, trifluoroacetic acid rhodium ii salt dimer, dirhodium ii tetra trifluoroacetate, rhodium ii trifluoroacetate dimer, ?rh2 cf3coo 4, bis 2,2,2-trifluoroacetyl oxy rhodio 2,2,2-trifluoroacetate
PubChem CID 10985187
IUPAC Name rhodium;2,2,2-trifluoroacetic acid
SMILES C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.[Rh].[Rh]

Specifications

Color Green
Physical Form Crystalline Powder
Quantity 250 mg
Formula Weight 657.88
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Rhodium(II) trifluoroacetate dimer
compliance-icons
Product Identifier
  • Rhodium(II) trifluoroacetate dimer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.