Learn More
Rhodium(III) 2,4-pentanedionate, Premion™, 99.99% (metals basis), Rh 25.2% min, Thermo Scientific Chemicals
Description
Rhodium(III) 2,4-pentanedionate is used in catalytic reagents for organic synthesis. Rhodium(III) 2,4-pentanedionate is useful for hydrosilylation of a variety of organic substrates like alkenes, terminal or internal acetylenes, conjugated dienes, alpha, beta -unsaturated aldehydes and allylic compounds. It performs the role of a catalyst in well-known reactions such as transformation processes including hydrogenation, ethanol steam reforming, CO oxidation, and NOx reduction. Rhodium acetylacetonate, as the organic solvent-soluble source of rhodium metal ions, can be used to prepare monodisperse size/shape controlled Rh nanocrystals. Rh nanocrystal monolayers have been reported to show high activity for hydrogenation of vinylcyclohexene to selectively produce ethylcyclohexene. The size/shape controlled nanocrystals find a variety of applications in oil refining processes, chemical synthesis and fuel cell technology.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 263°C to 264°C |
Physical Form | Crystalline |
Quantity | 0.5 g |
Solubility Information | Very soluble in chloroform; Soluble in alcohol; In soluble in water. |
IUPAC Name | rhodium(3+) tris((2Z)-4-oxopent-2-en-2-olate) |
Formula Weight | 400.23 |
Percent Purity | 99.99% |
Grade | Premion™ |
Concentration or Composition (by Analyte or Components) | Rh 25.2% min. |
Assay | (metals basis) |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W17K059 | Certificate of Analysis | 27/09/2024 | 010561.02,010561.06,010561.03 |
S19J007 | Certificate of Analysis | 23/06/2023 | 010561,010561.02,010561.06,010561.03 |
W08I004 | Certificate of Analysis | 14/09/2022 | 010561.02,010560.01,010560.03,010560.06 |
F10P33 | Certificate of Analysis | 08/12/2021 | 010561.02,010571.22,010561.06,010561.03 |
J31N28 | Certificate of Analysis | 08/12/2021 | 010561.02,010560.01,010560.03,010560.06 |
Safety and Handling


- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H361-Suspected of damaging fertility or the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.