missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) chloride, anhydrous, 48.6-49.2% Rh, Thermo Scientific Chemicals

Product Code. 10558902
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-07-7
Molecular Formula Cl3Rh
Molecular Weight (g/mol) 209.26
MDL Number MFCD00011204
InChI Key SONJTKJMTWTJCT-UHFFFAOYSA-K
IUPAC Name rhodium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Rh+3]

Specifications

Boiling Point 717.0°C (730.0 mmHg)
Melting Point 450.0°C
Color Black
Physical Form Crystalline Powder
Quantity 500 mg
Linear Formula RhCl3
Fieser 02,357; 03,242; 06,504; 07,313; 08,435; 11,460; 14,271; 16,292; 17,366
Merck Index 15, 8313
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkaline solution
Formula Weight 209.26
Percent Purity 48.6 to 49.2% Rh
Packaging Glass bottle
Specific Gravity 5.38
Density 5.3800g/mL
Chemical Name or Material Rhodium(III) chloride, 48.6-49.2% Rh
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Rhodium(III) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Germ cell mutagenicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.,Harmful if swallowed.
  • H290-May be corrosive to metals.,May be corrosive to metals.
  • H341-Suspected of causing genetic defects.
  • H410-Very toxic to aquatic life with long lasting effects.
Sicherheitshinweis
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.,IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/physician.,Immediately call a POISON CENTER/doctor/physician.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'