missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) chloride, anhydrous, 48.6-49.2% Rh

Product Code. p-41417 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
500mg
This item is not returnable. View return policy

Product Code. 10558902

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10049-07-7 | Cl3Rh | 209.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-07-7
Molecular Formula Cl3Rh
Molecular Weight (g/mol) 209.26
MDL Number MFCD00011204
InChI Key SONJTKJMTWTJCT-UHFFFAOYSA-K
IUPAC Name rhodium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Rh+3]

Specifications

Boiling Point 717.0°C (730.0 mmHg)
Melting Point 450.0°C
Color Black
Physical Form Crystalline Powder
Quantity 500 mg
Linear Formula RhCl3
Fieser 02,357; 03,242; 06,504; 07,313; 08,435; 11,460; 14,271; 16,292; 17,366
Merck Index 15, 8313
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkaline solution
Formula Weight 209.26
Percent Purity 48.6 to 49.2% Rh
Packaging Glass bottle
Specific Gravity 5.38
Density 5.3800g/mL
Chemical Name or Material Rhodium(III) chloride, 48.6-49.2% Rh
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Rhodium(III) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Germ cell mutagenicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.,Harmful if swallowed.
  • H290-May be corrosive to metals.,May be corrosive to metals.
  • H341-Suspected of causing genetic defects.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.,IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/physician.,Immediately call a POISON CENTER/doctor/physician.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.