Learn More
Rhodium(III) chloride, anhydrous, 99.9% (metals basis), Rh 48.7% min
Description
It is used in homogeneous catalysis, for example in the industrial prodcution of acetic acid. It is used to coat optic fibres and optical mirrors, and for crucibles, thermocouple elements and headlight reflectors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Spezifikation
| Color | Red to Brown |
| Physical Form | Powder |
| Quantity | 1 g |
| Linear Formula | RhCl3 |
| Merck Index | 14,8188 |
| Solubility Information | Insoluble in water. Soluble in hydroxide and cyanide solutions. |
| Formula Weight | 209.26 |
| Percent Purity | 99.9% |
| Concentration or Composition (by Analyte or Components) | Rh 48.7% min. |
| Odor | Odorless |
| Mehr anzeigen |
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.