missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) chloride, anhydrous, 99.9% (metals basis), Rh 48.7% min

Product Code. p-7048874
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11361858

Brand: Thermo Scientific Alfa Aesar 011815.01

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10049-07-7 | Cl3Rh | 209.26 g/mol

It is used in homogeneous catalysis, for example in the industrial prodcution of acetic acid. It is used to coat optic fibres and optical mirrors, and for crucibles, thermocouple elements and headlight reflectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-07-7
Molecular Formula Cl3Rh
Molecular Weight (g/mol) 209.26
MDL Number MFCD00011204
InChI Key SONJTKJMTWTJCT-UHFFFAOYSA-K
IUPAC Name rhodium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Rh+3]

Specifications

Color Red to Brown
Physical Form Powder
Quantity 0.25 g
Linear Formula RhCl3
Merck Index 14,8188
Solubility Information Insoluble in water. Soluble in hydroxide and cyanide solutions.
Formula Weight 209.26
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Rh 48.7% min.
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Rhodium(III) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.