missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodium(III) chloride hydrate, 38% Rh, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 100.0°C |
Color | Brown to Brown-Red |
Physical Form | Crystalline Powder |
Quantity | 5 g |
Linear Formula | RhCl3·xH2O |
Fieser | 10,343; 11,460; 14,271; 16,292; 17,366 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alchol and hcl, insoluble in ether |
Formula Weight | 209.26 |
Percent Purity | 38% (Rh) |
Packaging | Glass bottle |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0446844 | Certificate of Analysis | 02/11/2023 | 195520010,195520050 |
A0457091 | Certificate of Analysis | 25/10/2023 | 195520010,195520050 |
A0455494 | Certificate of Analysis | 25/08/2023 | 195520010,195520050 |
A0398392 | Certificate of Analysis | 14/12/2022 | 195520010,195520050 |
A0407636 | Certificate of Analysis | 12/12/2022 | 195520010,195520050 |
Safety and Handling




Product Identifier
- Rhodium(III) chloride hydrate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Germ cell mutagenicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- H318-Causes serious eye damage.
- H341-Suspected of causing genetic defects.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- H410-Very toxic to aquatic life with long lasting effects.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.,IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.,Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Rhodium(III) chloride hydrate, 38% Rh, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction