missing translation for 'onlineSavingsMsg'
Learn More
Rhodium(III) chloride hydrate, 99.95%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10803781

Rhodium(III) chloride hydrate, 99.95%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10803781
1 g, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10803781

Brand: Thermo Scientific Acros 437090010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20765-98-4
Molecular Formula Cl3Rh
Molecular Weight (g/mol) 209.26
MDL Number MFCD00149839
InChI Key SONJTKJMTWTJCT-UHFFFAOYSA-K
IUPAC Name rhodium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Rh+3]

Specifications

Melting Point 100.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 37.0 to 43% (Rh)
Linear Formula RHCl3·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alchol and hcl, insoluble in ether
Formula Weight 209.26
Percent Purity 99.95%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Rhodium(III) chloride hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460215 Certificate TypeCertificate of Analysis Date14/02/2024 Catalog Number437092500,437090010
Lot NumberA0443716 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number437092500,437090010
Lot NumberA0398329 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number437092500,437090010
Lot NumberA0396934 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number437092500,437090010
Lot NumberA0429731 Certificate TypeCertificate of Analysis Date11/05/2021 Catalog Number437092500,437090010
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Rhodium(III) chloride hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Germ cell mutagenicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.,IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/physician.,Immediately call a POISON CENTER/doctor/physician.,Immediately call a POISON CENTER/doctor/physician.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhodium(III) chloride hydrate, 99.95%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.