missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) iodide, 99.9% (metals basis), Rh 20.8% min

Product Code. 11361648
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11361648

Brand: Thermo Scientific Alfa Aesar 011029.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15492-38-3 | I3Rh | 483.62 g/mol

Rhodium(III) iodide is applied for carbonylation process. It is also used as a precursor for hydroformylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15492-38-3
Molecular Formula I3Rh
Molecular Weight (g/mol) 483.62
MDL Number MFCD00016311
InChI Key KXAHUXSHRWNTOD-UHFFFAOYSA-K
IUPAC Name rhodium(3+) triiodide
SMILES [Rh+3].[I-].[I-].[I-]

Specifications

Color Black
Physical Form Crystalline
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 483.62
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Rh 20.8% min.
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Rhodium(III) iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.