missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) iodide, 99.9% (metals basis), Rh 20.8% min, Thermo Scientific Chemicals

£126.63 - £2255.22

Chemical Identifiers

CAS 15492-38-3
Molecular Formula I3Rh
Molecular Weight (g/mol) 483.62
MDL Number MFCD00016311
InChI Key KXAHUXSHRWNTOD-UHFFFAOYSA-K
IUPAC Name rhodium(3+) triiodide
SMILES [Rh+3].[I-].[I-].[I-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11351648
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
011029.01
0.25 g
£126.63
0.25g
Please to purchase this item. Need a web account? Register with us today!
11341648
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
011029.03
1 g
£506.52
1g
Please to purchase this item. Need a web account? Register with us today!
11361648
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
011029.06
5 g
£2255.22
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rhodium(III) iodide is applied for carbonylation process. It is also used as a precursor for hydroformylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15492-38-3
483.62
KXAHUXSHRWNTOD-UHFFFAOYSA-K
[Rh+3].[I-].[I-].[I-]
I3Rh
MFCD00016311
rhodium(3+) triiodide

Specifications

15492-38-3
Crystalline
MFCD00016311
KXAHUXSHRWNTOD-UHFFFAOYSA-K
rhodium(3+) triiodide
483.62
Rh 20.8% min.
Hygroscopic
Rhodium(III) iodide
Black
I3Rh
Insoluble in water.
[Rh+3].[I-].[I-].[I-]
483.62
99.9%
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 239-521-5

missing translation for 'rtecsNumber' : VI9315000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.