Learn More
Rhodium(III) oxide pentahydrate, Premion™, 99.99% (metals basis), Rh 59% min, Thermo Scientific Chemicals
Description
Used as a catalytic convertor in automobiles. It is also employed as an industrial catalyst in the manufacturing of chemical intermediates such as oxo-alcohols, as well as nitric acid and ethanoic (acetic) acid productions. It is, likewise, of interest for new capacitor electrode materials, dynamic random access memories (DRAMs) and non-volatile random access memories (FeRAMs).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | (decomposition) |
Physical Form | Powder |
Quantity | 5 g |
Solubility Information | Insoluble in water and aqua regia. |
Formula Weight | 343.88 (253.81 Anhydrous) |
Percent Purity | 99.99% |
Grade | Premion™ |
Concentration or Composition (by Analyte or Components) | Rh 59% min. |
Assay | (metals basis) |
Chemical Name or Material | Rhodium(III) oxide pentahydrate |
Product Suggestions
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N04I006 | Certificate of Analysis | 22/11/2022 | 012667.06,012667.03,012667.01 |
N12F011 | Certificate of Analysis | 22/11/2022 | 012667.06,012667.03,012667.01 |
S14H029 | Certificate of Analysis | 22/11/2022 | 012667.06,012667.03,012667.01 |
N04I010 | Certificate of Analysis | 22/11/2022 | 012667.06,012667.03,012667.01 |
N04I006 | Certificate of Analysis | 12/04/2022 | 012667.06,012667.03,012667.01,012678.03 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.