Learn More
Rhodium(III) sulfate tetrahydrate, Premion™, 99.99% (metals basis), Rh 35.9% min, Thermo Scientific Chemicals
£208.00 - £3421.00
Chemical Identifiers
CAS | 15274-78-9 |
---|---|
Molecular Formula | H8O16Rh2S3 |
Molecular Weight (g/mol) | 566.04 |
MDL Number | MFCD00798544 |
InChI Key | JJITWKYISWZJBI-UHFFFAOYSA-H |
Synonym | rhodium iii sulfate tetrahydrate, 3so4.2rh.4h2o, rhodium iii sulfate tetrahydrate, premion, dirhodium 3+ tetrahydrate trisulfate |
PubChem CID | 91886287 |
IUPAC Name | rhodium(3+);trisulfate;tetrahydrate |
SMILES | O.O.O.O.[Rh+3].[Rh+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11379417
|
Thermo Scientific Alfa Aesar
041016.01 |
0.25 g |
£208.00
0.25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389417
|
Thermo Scientific Alfa Aesar
041016.03 |
1 g |
£825.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399417
|
Thermo Scientific Alfa Aesar
041016.06 |
5 g |
£3421.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rhodium(III) sulfate is used as plating reagents and additives for noble metal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15274-78-9 | |
566.04 | |
JJITWKYISWZJBI-UHFFFAOYSA-H | |
91886287 | |
O.O.O.O.[Rh+3].[Rh+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
H8O16Rh2S3 | |
MFCD00798544 | |
rhodium iii sulfate tetrahydrate, 3so4.2rh.4h2o, rhodium iii sulfate tetrahydrate, premion, dirhodium 3+ tetrahydrate trisulfate | |
rhodium(3+);trisulfate;tetrahydrate |
Specifications
15274-78-9 | |
Crystalline | |
Rh2(SO4)3·4H2O | |
UN3261 | |
Slightly soluble in water. | |
O.O.O.O.[Rh+3].[Rh+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
566.04 | |
566.05 (493.99 Anhydrous) | |
Premion™ | |
Odorless | |
Rhodium(III) sulfate tetrahydrate |
Brown | |
H8O16Rh2S3 | |
MFCD00798544 | |
rhodium iii sulfate tetrahydrate, 3so4.2rh.4h2o, rhodium iii sulfate tetrahydrate, premion, dirhodium 3+ tetrahydrate trisulfate | |
JJITWKYISWZJBI-UHFFFAOYSA-H | |
rhodium(3+);trisulfate;tetrahydrate | |
91886287 | |
99.99% | |
Rh 35.9% min. | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H318-H290
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 234-014-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only