missing translation for 'onlineSavingsMsg'
Learn More
Rhodium powder, -22 mesh, Premion™, 99.97% (metals basis), precious metals typically <20ppm each, Thermo Scientific™ Product Code.: 15484735

Rhodium powder, -22 mesh, Premion™, 99.97% (metals basis), precious metals typically <20ppm each, Thermo Scientific™

Product Code. 15484735
0.25 g
Click to view available options
Quantity:
0.25 g
This item is not returnable. View return policy

Product Code. 15484735

Brand: Thermo Scientific Alfa Aesar 010668.01

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]

Specifications

Color Gray
Physical Form Powder
Quantity 0.25 g
UN Number UN3089
Percent Purity 99.97%
Grade Premion™
Odor Odorless
Assay (metals basis)
Chemical Name or Material Rhodium powder, Precious metals typically <20ppm each

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS11G003 Certificate TypeCertificate of Analysis Date03/02/2023 Catalog Number010652.06,010668.01,010652.22,010652.14
Lot Number20992 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010652.06,010668.01,010652.22,010652.14
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhodium powder, -22 mesh, Premion™, 99.97% (metals basis), precious metals typically <20ppm each, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.