missing translation for 'onlineSavingsMsg'
Learn More
Rhodium sponge, -20 mesh, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 15471837

Rhodium sponge, -20 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 15471837
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15471837

Brand: Thermo Scientific Alfa Aesar 042866.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-16-6
Molecular Formula Rh
MDL Number MFCD00011201 MFCD03458392
UN Number UN3089
Quantity 1 g
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
SMILES [Rh]
IUPAC Name rhodium
Molecular Weight (g/mol) 102.91
PubChem CID 23948
ChEBI CHEBI:33359
Percent Purity 99.9%
Mesh Size −20 Mesh
Odor Odorless
Form Sponge
Assay (metals basis)
Chemical Name or Material Rhodium sponge
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhodium sponge, -20 mesh, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.