missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodizonic acid, disodium salt, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
Melting Point | >300.0°C |
Color | Gray-Green to Green |
Physical Form | Fine Crystalline Powder |
Quantity | 5 g |
Assay Percent Range | 98% |
Beilstein | 08,536 |
Merck Index | 15,8799 |
Solubility Information | (0.2% in water) Clear |
IUPAC Name | disodium;3,4,5,6-tetraoxocyclohexene-1,2-diolate |
Formula Weight | 214.04 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'