missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Ribbon Ink Cartridge
For use with Mettler Toledo pH meters and printers.
Brand: Mettler Toledo™ 65975
34.78 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
- Black ink
- Contains two ribbons

Specifications
Printers, pH Meter | |
Black | |
2 Ribbons/Pk. | |
Ribbon Cartridge |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mettler Toledo™ Ribbon Ink Cartridge > Ribbon Cartridge
Spot an opportunity for improvement?Share a Content Correction