missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ RiboLock RNase Inhibitor (40 U/μL)
Shop All Thermo Scientific Products
Click to view available options
Quantity:
2,500 units
24 x 2,500 units
4 x 2,500 units
Unit Size:
2500 units
Pack of 24
Pack of 4
Description
RiboLock RNase Inhibitor inhibits the activity of RNases A , B, and C by binding them in a noncompetitive mode at a 1:1 ratio. It does not inhibit eukaryotic RNases T1, T2, U1, U2, CL3 as well as prokaryotic RNases I and H.
Highlights
- Performs under a wide range of reaction conditions
- Protects RNA from degradation at temperatures up to 55°C
Specifications
Specifications
| Concentration | 40 U/μL |
| Compatible Buffer | Storage Buffer |
| Quantity | 24 x 2,500 units |
| Product Type | RNase Inhibitor |
| Product Line | RiboLock |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction