Learn More
Epredia™ Base Mold Release
Coat base molds for easy separation of paraffin from mold with Epredia™ Base Mold Release.
Brand: Epredia™ 6600
Additional Details : CAS Number : 67-63-0 Weight : 0.10000kg Transport : UN number : 1219 Chem class : 3 Pack group : II
Description
- Paraffin buildup on the molds is reduced
- Environmentally safe product is ready-to-use
- Non-aerosol pump spray bottle
Specifications
Base Mold Release | |
4 oz. |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H371-May cause damage to organs.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P235-Keep cool.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methyl alcohol, polyethylene glycol, isopropyl alcohol, polyehtylene glycol octylphenyl ether, ethyl