missing translation for 'onlineSavingsMsg'
Learn More
Epredia™ Cytoseal™ Mountant Product Code.: 11932365
  • ONLINE EXCLUSIVE PRICE
    £341.00
    £59.84 / Pack of 12
    Save £281.16
    82% Off
compliance-icons compliance-icons compliance-icons

Epredia™ Cytoseal™ Mountant

Product Code. p-4532396 Shop All Epredia Products
16 oz., Cytoseal 60, Pack of 12
Click to view available options
Quantity:
4 oz.
16 oz.
Product Line:
Cytoseal 60
Cytoseal XYL
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 11932365

Brand: Epredia™ 831016

  • ONLINE EXCLUSIVE PRICE
    £341.00
    £59.84 / Pack of 12
    Save £281.16
    82% Off
View Available Promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prevent cracking, fading and discoloration of stained specimens with Cytoseal™ mounting media.

  • Cytoseal 60 mountant is a low-viscosity medium that dries quickly and allows for rapid, even spreading which virtually eliminates air bubbles

Ordering Information:

Packaged in an easy-to-use dispenser with a twist-open/twist-closed cap.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Cytoseal 60
Product Type Mountant
Quantity 16 oz.
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H360-May damage fertility or the unborn child.
  • H370-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P235-Keep cool.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P363-Wash contaminated clothing before reuse.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : toluene, acrylic resin, butyl benzyl phthalate, 2,6-ditertbutyl-p-cresol
Promo
Expires: 2025-12-14
Save Up to 63%
Big savings on advanced technologies and materials testing. Promo Code: 24141.

Applying Promo and Adding...

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ Cytoseal™ Mountant >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.