Learn More
Epredia™ Decalcifying Solution
Remove calcium from histological specimens with Epredia™ Decalcifying Solution.
Brand: Epredia™ 8340
This item is not returnable.
View return policy
Description
- Offers rapid decalcification without destroying nuclear properties of the tissue
- The EDTA and dilute hydrochloric acid in this solution require no heat, electricity, or other aids to decalcify bone tissue
Specifications
Decalcifying Solution | |
32 oz. |
Product Content Correction
Product Title
Epredia™ Decalcifying Solution > 32 oz.
Spot an opportunity for improvement?Share a Content Correction