Learn More
Epredia™ Decalcifying Solution
Product Code. p-4532363 Shop All Epredia Products
Click to view available options
Quantity:
32 oz.
Unit Size:
Pack of 6
Product Code. 12697766
Brand: Epredia™ 8340
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Remove calcium from histological specimens with Epredia™ Decalcifying Solution.
  • Offers rapid decalcification without destroying nuclear properties of the tissue
  • The EDTA and dilute hydrochloric acid in this solution require no heat, electricity, or other aids to decalcify bone tissue
TRUSTED_SUSTAINABILITY
Specifications
Product Type Decalcifying Solution
Quantity 32 oz.