Learn More
Epredia™ Richard-Allan Scientific™ Eosin-Y Alcoholic, Eosin-Y with Phloxine, Eosin-Y Saturated, Eosin-Y Alcoholic
Enjoy strict quality control of raw materials and finished product and a high quality product every time with the Epredia™ Richard-Allan Scientific™ Eosin-Y Alcoholic, Eosin-Y with Phloxine, and Eosin-Y Saturated.
Brand:  Epredia™ 71211
Code : LE
Additional Details : CAS Number : 64-17-5 Weight : 4.08237kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
 View more versions of this product
Product Code. 12697806
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Strict quality control of raw materials and finished product ensures a high quality product every time
  • Eosin-Y Alcoholic produces three distinct color variations enhancing differentiation between muscle, red blood cells, and connective tissue
  • Eosin-Y w/Phloxine yields a brighter reddish hue and a more dramatic differentiation of cytoplasmic color than Eosin-Y Alcoholic
  • Eosin-Y Saturated is a very intense and rapid counterstain that can be used to make working solutions of Eosin-Y
Specifications
Specifications
Alcoholic Eosin Y Stain
1 gal.