Description
Features:- Easy to clean
- Chemical resistant
- Autoclavable
- Choose the correct tool to gross specimens at 3, 4 or 5mm thickness.
- Includes hex wrench and a 10-pack of high profile disposable blades.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Epredia™ Richard-Allan Scientific™ Fine Cutting Tool Kit, 5mm