Learn More
Epredia™ Richard-Allan Scientific™ Fix-Rite™ 2, Pump Spray Bottle
Perform routine cytological preparations with Epredia™ Richard-Allan Scientific™ Fix-Rite™ 2.
Brand:  Epredia™ 76150
Product Code. 12592726
  • £181.00 / Pack of 12
Explore more special offers
Description
Description
  • Coating protects cells when mailing or transporting slides to the laboratory
  • Packaged in 4 oz. containers to allowing complete control during application
Specifications
Specifications
Fixative