missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Mark-It™ Tissue Marking Dyes

For a variety of tissue identification purposes use Epredia™ Mark-It™ Tissue Marking Dyes.

Brand:  Epredia™ 5000O


 View more versions of this product

Product Code. 12628076

  • £77.65 / Each
Estimated Shipment: 04-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Features:
  • Permanent dyes that maintain their original brilliance during routine tissue processing cycles
  • Can be used on fresh or formalin-fixed tissues

Uses include:

  • Marking surgical margins
  • Identifying serial segments of large tissues to ensure proper orientation and embedding
  • Maintaining proper tissue orientation for Mohs surgery specimens
  • Identification of multiple pieces of tissues processed in the same cassette

Ordering Information:

  • Available in black, blue, green, orange, red and yellow in 2 oz. bottles
Specifications

Specifications

Orange
Tissue Marking Dye
2 oz./ ea.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P362-Take off contaminated clothing.

Supplemental information
  • MIXTURE LIST-Contains : ammonium hydroxide,butanamide, 2,2'-[(3,3'-dimethoxy[1,1'-biphenyl]-4,4'-diyl)bis(azo)]Bis[3-oxo-N-

SDS
Documents

Documents

Certificates
Special Offers

Special Offers